Learn More
Oxidation Reagent,Solstice
Product Code. 16865421
Change view
Click to view available options
4X2.5L GLASS BOTTLE
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
16865421 4X2.5L GLASS BOTTLE
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16865421 Supplier Fluka Supplier No. BR6634X2.5L
  • ONLINE EXCLUSIVE PRICE
    2505.00 EUR
    1036.35 EUR / Pack of 4
    Save 1468.65 EUR
    58% Off
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products
BioSyn™, suitable for use with DNA/RNA synthesis instruments, 0.02M Iodine in 78/20/2 THF/Pyridine/Water (v/v)
Solstice BioSyn™ reagents and solvents for DNA/RNA synthesis
  • Solstice BioSyn™ Solvents and Reagents are specifically processed and purified to exacting specifications, ensuring outstanding synthesis efficiency
  • ideal for oligonucleotide synthesis regardless of the application
  • All solvents and reagents are filtered (0.2-micron for oxidation reagents and 0.1-micron for all other reagents) to minimize particulate contamination
TRUSTED_SUSTAINABILITY
Specifications
CAS 109-99-9, 110-86-1, 7732-18-5, 7553-56-2
CAS Max % 78.00%, 20.00%, 2.00%, 0.20%
Quantity 4 x 2.5 L
Flash Point -15°C
Packaging Glass Bottle
Physical Form Liquid, clear
Purity Grade Notes Oxidation reagents
Boiling Point 66°C