Learn More
Oxatomide, TRC
Product Code. 30426626
Change view
Click to view available options
Quantity:
5 mg
10 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30426626 100 mg
30417418 5 mg
30414244 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30426626 Supplier Toronto Research Chemicals Supplier No. TRCO845100100MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 60607-34-3
Molecular Formula C27 H30 N4 O
Molecular Weight (g/mol) 426.55
Synonym 2H-Benzimidazol-2-one, 1-[3-[4-(diphenylmethyl)-1-piperazinyl]propyl]-1,3-dihydro-, Celtect, Cobiona, Dasten, KW 4354, NSC 309710, Oxatomide, R 35443, Tinset
IUPAC Name 3-[3-(4-benzhydrylpiperazin-1-yl)propyl]-1H-benzimidazol-2-one
SMILES O=C1Nc2ccccc2N1CCCN3CCN(CC3)C(c4ccccc4)c5ccccc5
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C27H30N4O/c32-27-28-24-14-7-8-15-25(24)31(27)17-9-16-29-18-20-30(21-19-29)26(22-10-3-1-4-11-22)23-12-5-2-6-13-23/h1-8,10-15,26H,9,16-21H2,(H,28,32)
Formula Weight 426.242
Chemical Name or Material Oxatomide