Learn More
Oxatomide, Mikromol™
Product Code. 17982496
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17982496 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17982496 Supplier Mikromol Supplier No. MM1115.00
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Oxatomide
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Oxatomide
CAS 60607-34-3
Analyte or Component Names Oxatomide
Shipping Condition Room Temperature
Molecular Formula C27 H30 N4 O
Certifications/Compliance ISO 17025
Synonym 2H-Benzimidazol-2-one, 1-[3-[4-(diphenylmethyl)-1-piperazinyl]propyl]-1,3-dihydro-, Celtect, Cobiona, Dasten, KW 4354, NSC 309710, Oxatomide, R 35443, Tinset
InChI Formula InChI=1S/C27H30N4O/c32-27-28-24-14-7-8-15-25(24)31(27)17-9-16-29-18-20-30(21-19-29)26(22-10-3-1-4-11-22)23-12-5-2-6-13-23/h1-8,10-15,26H,9,16-21H2,(H,28,32)
SMILES O=C1Nc2ccccc2N1CCCN3CCN(CC3)C(c4ccccc4)c5ccccc5
IUPAC Name 3-[3-(4-benzhydrylpiperazin-1-yl)propyl]-1H-benzimidazol-2-one
Molecular Weight (g/mol) 426.55
Quantity 250 mg
Formula Weight 426.242 g/mol