Learn More
Oxatomide, LoGiCal
Product Code. 30461640
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30461640 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30461640 Supplier LoGiCal Supplier No. LGCFOR1115.00
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Oxatomide
CAS 60607-34-3
Analyte or Component Names Oxatomide
Shipping Condition Room Temperature
Molecular Formula C27 H30 N4 O
Certifications/Compliance ISO 9001
Synonym 2H-Benzimidazol-2-one, 1-[3-[4-(diphenylmethyl)-1-piperazinyl]propyl]-1,3-dihydro-, Celtect, Cobiona, Dasten, KW 4354, NSC 309710, Oxatomide, R 35443, Tinset
InChI Formula InChI=1S/C27H30N4O/c32-27-28-24-14-7-8-15-25(24)31(27)17-9-16-29-18-20-30(21-19-29)26(22-10-3-1-4-11-22)23-12-5-2-6-13-23/h1-8,10-15,26H,9,16-21H2,(H,28,32)
Solution Type Neat
SMILES O=C1Nc2ccccc2N1CCCN3CCN(CC3)C(c4ccccc4)c5ccccc5
IUPAC Name 3-[3-(4-benzhydrylpiperazin-1-yl)propyl]-1H-benzimidazol-2-one
Industry Type Clinical Testing
Molecular Weight (g/mol) 426.55
Quantity 10 mg
Formula Weight 426.242 g/mol