Learn More
Oxamyl-d6, TRC
Specifications
Specifications
| Chemical Name or Material | Oxamyl-d6 |
| CAS | 2470229-60-6 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C7H7D6N3O3S |
| Quantity | 10 mg |
| Synonym | 2-(Dimethylamino)-N-[[(methylamino)carbonyl]oxy]-2-oxo-ethanimidothioic Acid Methyl Ester-d6, N',N'-Dimethyl-N-[(methylcarbamoyl)oxy]-1-thio-Oxamimidic Acid Methyl Ester-d6, Methylcarbamic Acid O-[[[(dimethylcarbamoyl)methylthio]methylene]amino] deriv.-d6, DPX 1410-d6, DPX 1410L-d6, Du Pont 1410-d6, NSC 379588-d6, Oxamyl (pesticide)-d6, Thioxamyl-d6, Vydate-d6, Vydate CLV-d6, Vydate G-d6, Vydate L-d6 |
| InChI Formula | InChI=1S/C7H13N3O3S/c1-8-7(12)13-9-5(14-4)6(11)10(2)3/h1-4H3,(H,8,12)/b9-5+/i2D3,3D3 |
| SMILES | O=C(/C(SC)=N/OC(NC)=O)N(C([2H])([2H])[2H])C([2H])([2H])[2H] |
| Molecular Weight (g/mol) | 225.3 |
| Percent Purity | >95 |