Learn More
Thermo Scientific™ Owl™ EasyCast™ B1A Mini Gel and A2-OK Multi Gel System Replacement Parts
Product Code. 11962955 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Caster
Gasket
Gel Tray
Unit Size:
2 sets
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
11962955 Gel Tray Each
11942955 Caster Each
11952955 Gasket 2 sets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11962955 Supplier Thermo Scientific™ Supplier No. B1AUVT
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
Replace your UVT gel tray, gaskets and external casters with mini gel and multi gel system replacement parts.
Replace your UVT gel tray, gaskets and external casters with Thermo Scientific™ Owl™ EasyCast™ B1A Mini Gel and A2-OK Multi Gel System Replacement Parts, for use with EasyCast B1A and A2-OK Multi Gel Mini Gel Systems.
Ideal use with EasyCast B1A and A2-OK Multi Gel Mini Gel Systems.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Owl™ EasyCast B1A Mini Gel and A2-OK Multi Gel Electrophoresis System
Gel Size Mini
Product Line Owl, EasyCast
Type Gel Tray
Length (Metric) 8 cm
Width (Metric) 7 cm