Learn More
Thermo Scientific™ Owl™ EasyCast™ B1 Mini Gel Electrophoresis Systems
Product Code. 11942935
Change view
Click to view available options
Includes:
Buffer Exchange Ports
Without Buffer Exchange Ports
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Unit Size
11942935 Buffer Exchange Ports Each
11932935 Without Buffer Exchange Ports Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11942935 Supplier Thermo Scientific™ Supplier No. B1BP
  • ONLINE EXCLUSIVE PRICE
    870.00 EUR
    527.00 EUR / Pack of 1
    Save 343.00 EUR
    39% Off
View all promotions
In Stock
Cast and run a gel in the same chamber with no tape or additional parts required with the leakproof casting of mini gel electrophoresis systems.
Cast and run a gel in the same chamber with no tape or additional parts required with the leakproof casting of Thermo Scientific™ Owl™ EasyCast™ B1 Mini Gel Electrophoresis Systems. Durability and ease of use makes this the perfect choice for students and high volume researchers.
Features:
  • System offers a wide range of combs.
  • Two comb slots on gel tray double sample capacity by doubling number of sample wells.
  • UVT gel trays are silk screened with a florescent ruler for easy measurement of bands.
Includes:
  • Main buffer chamber
  • Thermo Scientific™ SuperSafe™ lid with attached power supply leads
  • Thermo Scientific™ EasyCast™ Gasketed UVT gel tray
  • Two Combs: 10 & 14 Well, Double-sided, 1.0/1.5mm thick
Ordering Information:
Available with optional buffer exchange ports, (B1-BP), for use with an external recirculation pump.
Warranty and Service Offering:
  • Three years
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance CE marked
Gel Size Mini
Product Line Owl
Type Mini Gel Electrophoresis System
Includes Buffer Exchange Ports
Sample Size 5 to 34
Volume (Metric) Buffer 600 mL
Dimensions (L x W x H) 8.63 x 5.88 x 3.75 in. (22 x 15 x 9.5 cm)
Length (English) 8.63 in.
Length (Metric) 22 cm
Length (Metric) Gel 11 cm
Width (English) 5.88 in.
Width (Metric) 15 cm
Width (Metric) Gel 9 cm
Height (English) 3.75 in.
Height (Metric) 9.5 cm