Learn More
Thermo Scientific™ Owl™ Dual-Gel Vertical Electrophoresis Systems
Product Code. 12474677
Change view
Click to view available options
Volume (Metric) Buffer:
300 mL
550 mL
1250 mL
Includes:
Apparatus Only
Complete System
Complete System plus Caster
Unit Size:
Each
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Buffer Includes Unit Size
12474677 1250 mL Apparatus Only Each
11903015 1250 mL Complete System plus Caster Each
11913015 1250 mL Complete System Each
11913005 300 mL Complete System plus Caster Each
11923005 300 mL Complete System Each
12404707 300 mL Apparatus Only Each
11973005 550 mL Complete System plus Caster Each
11983005 550 mL Complete System Each
11993005 550 mL Apparatus Only Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 12474677 Supplier Thermo Scientific™ Supplier No. P10DS3
  • ONLINE EXCLUSIVE PRICE
    1580.00 EUR
    1512.00 EUR / Pack of 1
    Save 68.00 EUR
    4% Off
Estimated Shipment: 10-02-2027
Ajouter au panier
Ajouter au panier
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Quickly and easily cast and run one or two gels in the same device with these Thermo Scientific™ Owl™ Dual-Gel Vertical Electrophoresis Systems.
Quickly and easily cast and run one or two gels in the same device using Thermo Scientific™ Owl™ Dual-Gel Vertical Electrophoresis Systems—comprised of the P8DS, P9DS and P10DS Dual Gel Systems.
Simple, Six-Step Use:
  1. Secure the glass cassettes
  2. Pour the agarose
  3. Plug in the specially designed trough
  4. Cast polyacrylamide solution in the cassettes
  5. Load samples
  6. Run the gel
Features:
  • Manufactured using a time-tested acrylic fabrication technology, providing a durable, long-lasting system
  • Central cooling core provides even cooling across the gel surface for outstanding resolution and clear banding
  • Cooling is achieved by circulating fluid through the core with external circulator or cool tap water
  • Very simple clamping mechanism makes loading glass cassettes and most manufacturers' precast gels trouble free
Recommended for:
  • SDS electrophoresis
  • The second dimension of two-dimensional (2-D) electrophoresis
  • Agarose electrophoresis
  • Native electrophoresis
TRUSTED_SUSTAINABILITY
Spécification
Product Line Owl
Type Dual-Gel Vertical Electrophoresis System Apparatus
Gel Dimensions 20 x 20 cm
Includes Apparatus Only
Model P10DS
Volume (Metric) Buffer 1250 mL
Length (English) 12 in.
Length (Metric) Gel 20 cm
Width (English) 6.5 in.
Width (Metric) Gel 20 cm
Height (English) 9 in.
Afficher plus Afficher moins
Are Bio-Rad power supplies compatible with Owl Electrophoresis systems?
The Owl Electrophoresis Systems are supplied with standard power leads, but we have not tested compatibility of the Owl systems with the Bio-Rad power supplies. For best performance, we recommend using one of our Owl or PowerEase power supplies for the Owl Electrophoresis Systems.
Nom du produit
Sélectionnez un problème
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.