Learn More
Honeywell™ Overspec Safety Glasses
Description
- Over-the-glass coverage, allowing workers full protection while wearing prescription eyewear
- Adjustable and bendable
- Wire-core temples offer flexibility for a secure, customized fit
- Ratchetting temples (3 positions) to further customize fit
- For use in agriculture, manufacturing, and pharmaceutical industries
- Protection from abrasion, airborne particulates, biohazard, impact
Specifications
Specifications
| Certifications/Compliance | EC Category PPE: 2, QA: ISO 9001/2000, EC Certificate: C899.1B-D, EC Attestation: C899.3B-D |
| Lens Coating | Scratch-resistant |
| Lens Tint | Clear |
| For Use With (Application) | For use in agriculture, manufacturing, and pharmaceutical industries |
| Frame Color | Blue, Black |
| Size | Large |
| Lens Material | Polycarbonate |
| Frame Material | Nylon |