missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
OSR1 Polyclonal antibody specifically detects OSR1 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | OSR1 |
| Applications | ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Formulation | PBS (pH 7.3), 50% glycerol |
| Gene Alias | ODDodd-skipped (Drosophila) homolog, odd-skipped homolog, odd-skipped related 1 (Drosophila), protein odd-skipped-related 1 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 28-115 of human OSR1 (NP_660303.1).,, Sequence:, MGSKTLPAPVPIHPSLQLTNYSFLQAVNGLPTVPSDHLPNLYGFSALHAVHLHQWTLGYPAMHLPRSSFSKVPGTVSSLVDARFQLPAFPWFPHVIQPKPEITAGGSVPALKTKPRFDFANLALAATQEDPAKLGRGEGPGSPAGGLGALLDVTKLSPEKKPTRGRLPSKTKKEFVCKFCGRHFTKSYNLLIHERTHTDERPYTCDICHKAFRRQDHLRDHRYIHSKEKPFKCQECGKGFCQSRTLAVHKTLHSQVKELKTSKIKC |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?