missing translation for 'onlineSavingsMsg'
Learn More

Osimertinib mesylate, MedChemExpress

Product Code. 16491196
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
500 mg
1 g
Unit Size:
100mg
10mg
1g
200mg
500mg
50mg
5mg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16491196 200 mg 200mg
16486926 100 mg 100mg
16482887 50 mg 50mg
16466818 10 mg 10mg
16451786 500 mg 500mg
16419367 5 mg 5mg
16411559 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 16491196 Supplier Medchem Express Supplier No. HY15772A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Osimertinib mesylate (AZD9291 mesylate) is a covalent, orally active, irreversible, and mutant-selective EGFR inhibitor with an apparent IC50 of 12 nM against L858R and 1 nM against L858R/T790M. Osimertinib overcomes T790M-mediated resistance to EGFR inhibitors in lung cancer.

Osimertinib mesylate (AZD9291 mesylate) is a covalent, orally active, irreversible, and mutant-selective EGFR inhibitor with an apparent IC50 of 12 nM against L858R and 1 nM against L858R/T790M. Osimertinib overcomes T790M-mediated resistance to EGFR inhibitors in lung cancer.

Specifications

Chemical Name or Material Osimertinib mesylate
CAS 1421373-66-1
Color Yellow
Purity Grade Notes Research
Molecular Formula C29H37N7O5S
Quantity 200 mg
Synonym AZD-9291 mesylate Mereletinib mesylate
Solubility Information DMSO : 13.89 mg/mL (23.32 mM; ultrasonic and warming and heat to 80°C)
SMILES C=CC(NC1=CC(NC2=NC=CC(C3=CN(C)C4=C3C=CC=C4)=N2)=C(OC)C=C1N(CCN(C)C)C)=O.CS(=O)(O)=O
Molecular Weight (g/mol) 595.71
Formula Weight 595.71
Percent Purity 98.74%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.