Learn More
Ortho-Phosphoric Acid, MilliporeSigma
Product Code. 15833920
Change view
Click to view available options
Quantity:
0.5 L
1 L
2.5 L
Unit Size:
0.50 L
1 L
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15833920 1 L 1 L
15843920 2.5 L 2.5 L
32210735 0.5 L 0.50 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15833920 Supplier Supelco Supplier No. 1005731000
  • ONLINE EXCLUSIVE PRICE
    132.00 EUR
    85.12 EUR / 1 L
    Save 46.88 EUR
    35% Off
View all promotions
 Restricted Item
In Stock
85%, for analysis EMSURE™ ACS,ISO,Reag. Ph Eur
Concentrated phosphoric acid is approx. 85% H3PO4 (aqueous)
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7664-38-2
Molecular Weight (g/mol) 98 g/mol
Specifications
Packaging Plastic bottle
Quantity 1 L
Percent Purity 85%
Grade ACS
Chemical Name or Material Ortho-Phosphoric acid
compliance-icons compliance-icons
Product Identifier
  • ortho-Phosphoric acid 85% for analysis EMSURE® ACS,ISO,Reag. Ph Eur
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Acute toxicity Category 4
Hazard Statement
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P234-Keep only in original container.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.