missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Orphenadrine Citrate, Mikromol™
Description
Orphenadrine Citrate
Specifications
Specifications
| Chemical Name or Material | Orphenadrine citrate |
| CAS | 4682-36-4 |
| Analyte or Component Names | Orphenadrine citrate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H23 N O . C6 H8 O7 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Orphenadrine citrate, (RS)-N,N-Dimethyl-2-[(2-methylphenyl)phenylmethoxy]ethanamine dihydrogen 2-hydroxypropane-1,2,3-tricarboxylate |
| InChI Formula | InChI=1S/C18H23NO.C6H8O7/c1-15-9-7-8-12-17(15)18(20-14-13-19(2)3)16-10-5-4-6-11-16;7-3(8)1-6(13,5(11)12)2-4(9)10/h4-12,18H,13-14H2,1-3H3;13H,1-2H2,(H,7,8)(H,9,10)(H,11,12) |
| SMILES | CN(C)CCOC(c1ccccc1)c2ccccc2C.OC(=O)CC(O)(CC(=O)O)C(=O)O |
| IUPAC Name | N,N-dimethyl-2-[(2-methylphenyl)-phenylmethoxy]ethanamine;2-hydroxypropane-1,2,3-tricarboxylic acid |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?