Learn More
Thermo Scientific™ Orion™ Star Series RS232 to USB Cable Adapter
Product Code. 11742637
Change view
Click to view available options
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size
11742637 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11742637 Supplier Thermo Scientific™ Supplier No. 1010005
  • ONLINE EXCLUSIVE PRICE
    138.00 EUR
    131.00 EUR / Pack of 1
    Save 7.00 EUR
    5% Off
Estimated Shipment: 22-09-2026
Connect a USB port to the RS232 port of your meter using the Thermo Scientific™ Orion™ Star Series RS232 to USB Cable Adapter.
Connect a USB port to the RS232 port of your meter using the Thermo Scientific™ Orion™ Star Series RS232 to USB Cable Adapter. This adapter allows interface between a USB connection and Thermo Scientific™ Orion™ Versa Star meters, Thermo Scientific™ Orion™ Star A series (Star A320, Star A220 and Star A210 series) meters and legacy Orion™ Star and Star Plus meters.
TRUSTED_SUSTAINABILITY
Specifications
Description Orion Star series RS232 to USB adapter cable
Type Meter Interface Cable
Includes RS232 to USB computer interface adapter and cable (2.5mm RS232 meter connector and USB computer connector)