Learn More
Thermo Scientific™ Orion™ RDO Stainless Steel Probe Guard
Product Code. 11742607
Change view
Click to view available options
Type:
Probe Guard
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
11742607 Probe Guard Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11742607 Supplier Thermo Scientific™ Supplier No. 087002
  • ONLINE EXCLUSIVE PRICE
    455.00 EUR
    424.00 EUR / Pack of 1
    Save 31.00 EUR
    6% Off
Estimated Shipment: 16-11-2026
Use this stainless-steel probe guard with RDO probes.
Use the Thermo Scientific™ Orion™ RDO™ Stainless Steel Probe Guard with Orion RDO probes.
TRUSTED_SUSTAINABILITY
Specifications
Description Probe Guard
For Use With (Equipment) Orion RDO Probes 087010MD, 087020MD, 087030MD, 087050MD, 087100MD
Type Probe Guard
Material Stainless Steel