Learn More
Thermo Scientific™ Orion™ Gas-Sensing ISE membranes
Product Code. 10165673
Change view
Click to view available options
For Use With (Equipment):
Carbon Dioxide Electrode, 9502BNMP
HP Ammonia Electrode, 9512HPBNWP
HP Ammonia Electrode, 9512HPBNWP, 9512HP
Standard Ammonia Electrode, 9512BNWP, 9512
Includes:
20 Loose Membranes
3 Bonded Membranes
3 Membranes
4 Membranes
Unit Size:
Each
Pack of 20
Pack of 3
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Includes Unit Size
10165673 HP Ammonia Electrode, 9512HPBNWP 3 Membranes Pack of 3
10408532 Carbon Dioxide Electrode, 9502BNMP 4 Membranes Each
10292441 Standard Ammonia Electrode, 9512BNWP, 9512 20 Loose Membranes Pack of 20
10758971 Standard Ammonia Electrode, 9512BNWP, 9512 3 Bonded Membranes Pack of 3
10554454 HP Ammonia Electrode, 9512HPBNWP, 9512HP 20 Loose Membranes Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 10165673 Supplier Thermo Scientific™ Supplier No. 951215
  • ONLINE EXCLUSIVE PRICE
    90.90 EUR
    84.80 EUR / Pack of 3
    Save 6.10 EUR
    6% Off
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
Choose from loose or preassembled membranes for ease of use with Thermo Scientific™ Orion™ Gas-Sensing ISE membranes.
Choose from loose or preassembled membranes for ease of use with Thermo Scientific™ Orion™ Gas-Sensing ISE membranes.
Fresh membranes provide optimum performance with gas-sensing ion selective electrodes. Ammonia electrodes require different membranes if using standard electrode or high performance ammonia electrode.
TRUSTED_SUSTAINABILITY
Specifications
Description Gas-Sensing ISE membrane
For Use With (Equipment) HP Ammonia Electrode, 9512HPBNWP
Type Gas-Sensing ISE Membrane
Includes 3 Membranes