Learn More
Thermo Scientific™ Orion™ AquaMate Vis and UV-Vis Spectrophotometers
Product Code. p-7160112
Change view
Click to view available options
Type:
7100 Vis Spectrophotometer
8100 UV-Vis Spectrophotometer
Plug Type:
China, India, ANZ Power Cord
NA, EU, UK Power cord
Spectral Bandwidth:
2 nm
5 nm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Plug Type Spectral Bandwidth unitSize
17143841 7100 Vis Spectrophotometer NA, EU, UK Power cord 5 nm Each
16439835 8100 UV-Vis Spectrophotometer NA, EU, UK Power cord 2 nm Each
17314773 8100 UV-Vis Spectrophotometer China, India, ANZ Power Cord 2 nm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17143841 Supplier Thermo Scientific™ Supplier No. AQ7100
Beinhaltet
Test Tube Holder for vials and test tube from 12 to 25 mm OD, AquaMate Water Analysis Methods Software, Dust Cover, External Power Supply, North America power cable (NEMA 5-15), European power cable (CEE 7/7), UK Power cable (BS 1362)
Optimized for your water analysis needs AquaMate 7100 Vis and AquaMate 8100 UV-Vis Spectrophotometers provide fast and reliable results
Thermo Scientific™ Orion™ AquaMate UV-Vis spectrophotometers include over 260 preprogrammed methods, and are designed to meet the needs of water and wastewater analysis laboratories. AquaMate UV-Vis and Visible-only spectrophotometers incorporate advanced features and functions for ultimate flexibility in operation.
Pre-programmed Methods:
  • More than 260 preprogrammed methods offer easy and convenient measurements using Orion AQUAfast™, Merck™ or CHEMetrics™ chemistries
  • Preprogrammed methods can also be adapted for new chemistries or operators can create their own custom methods and calibration curves
  • Instruments allow a one-point adjustment on any preprogrammed method to correct for variations in batch reagent chemistries
Versatility:
  • Glove friendly 7-inch touch screen controller
  • Wide variety of vials sizes can be used, including 10mm square vials, 20 and 50mm rectangular vials and an adjustable round vial holder that accommodates vials and test tube from 12 to 25 mm OD
Verification:
  • Performance verification tests ensure wavelength accuracy and instrument functionality
  • Each verification report gives the time, date and instrument serial number in accordance with GLP and GMP
  • Built-in wavelength accuracy test is compatible with either the internal lamp or external calibrated standards
  • Built-in test methods for stray light, noise and resolution are available to validate instrument performance further
Compliance
CE / TUV
Garantie
1 Year
TRUSTED_SUSTAINABILITY
Spezifikation
Baseline Flatness ±0.005 A
Certifications/Compliance CE / TUV
Compartments Accessible from top, front or side Able to accommodate cells up to 100 mm cells Available accessory for test tubes up to 25 mm diameter and 150 mm height Removable, washable sample compartment liner with magnetic placement and hold-down
Connections Single USB-A
Export data to network or PC via USB, Ethernet, or Wi-Fi USB adaptor, available
Print via USB, Ethernet or Wi-Fi USB adaptor available
Depth (Metric) 35.5 cm
Detector Type Dual Silicon Photodiodes
Display 7 in. High Definition Color Touchscreen, Fixed, 800 x 1280 pixels
Dimensions (L x W x H) 35.5 x 38.5 x 19.5 cm
Light Source Tungsten Halogen
Spectral Bandwidth 5 nm
Type 7100 Vis Spectrophotometer
Wavelength Range 325 to 1100 nm
Drift <0.0010 A/hr. (At 500 nm after 1 hour warmup)
Electrical Requirements External AC to DC converter. Voltage and Frequency (Hz) selected automatically, 100/240 V, 50/60 Hz
For Use With (Application) Water and Waste Water Analysis
Height (Metric) 19.5 cm
Includes
Test Tube Holder for vials and test tube from 12 to 25 mm OD, AquaMate Water Analysis Methods Software, Dust Cover, External Power Supply, North America power cable (NEMA 5-15), European power cable (CEE 7/7), UK Power cable (BS 1362).
Interface Glove friendly touch-screen
Keypad Glove- friendly touchscreen
Lamp Life >1000 hrs.
Length (Metric) 35.5 cm
Noise (RMS at 500 nm 60 consecutive measurements) ≤0.00020A at 0A at 260 and 500 nm ≤0.00030A at 1A at 260 and 500 nm ≤0.00040A at 2A at 260 and 500 nm
Optical Design Double Beam
Photometric Accuracy ±0.002 A at 0.5 A, ±0.004 A at 1.0 A, ±0.008 A at 2.0 A
Photometric Display 3A to +5A
Photometric Range –3A to +3.5A
Photometric Repeatability ±0.001A at 1A measured at 1.0A at 546 nm
Plug Type NA, EU, UK Power cord
Printer Included Snap-on printer available
Scan Speed Automatic – up to 1800 nm/min.
Stray Light <0.05%T at 340 nm and 400 nm
Warranty 1 Year
Wavelength Accuracy ±0.5 nm
Wavelength Data Interval 0.2 nm, 0.5 nm, 1 nm, 2 nm, 5 nm
Wavelength Repeatability < ±0.2 nm
Weight (Metric) 7.5 kg
Width (Metric) 38.5 cm
Mehr anzeigen Weniger anzeigen