missing translation for 'onlineSavingsMsg'
Learn More

Oregon Pesticide Mixture 3 100x AL 100-200 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30324882
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30324882 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30324882 Supplier Dr. Ehrenstorfer Supplier No. DREGA09000257AL

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Acetamiprid∣Azoxystrobin∣Bifenazate∣Bifenthrin∣Carbaryl∣Carbofuran∣Chlorantraniliprole∣Chlorpyrifos∣Clofentezine∣Diazinon∣Dimethoate∣Ethoprophos∣Etoxazole∣Fenoxycarb∣Imazalil∣Malathion∣Metalaxyl∣Methiocarb∣MGK 264∣MGK 264 isomer A∣MGK 264 isomer B∣Myclobutanil∣Parathion-methyl∣Permethrin∣Phosmet∣Prallethrin∣Propoxur∣Pyridaben∣Spinosad∣Spiromesifen∣Spirotetramat∣Thiacloprid∣Thiamethoxam∣Trifloxystrobin
CAS 135410-20-7, 131860-33-8, 149877-41-8, 82657-04-3, 63-25-2, 1563-66-2
Analyte or Component Names Acetamiprid∣Azoxystrobin∣Bifenazate∣Bifenthrin∣Carbaryl∣Carbofuran∣Chlorantraniliprole∣Chlorpyrifos∣Clofentezine∣Diazinon∣Dimethoate∣Ethoprophos∣Etoxazole∣Fenoxycarb∣Imazalil∣Malathion∣Metalaxyl∣Methiocarb∣MGK 264∣MGK 264 isomer A∣MGK 264 isomer B∣Myclobutanil∣Parathion-methyl∣Permethrin∣Phosmet∣Prallethrin∣Propoxur∣Pyridaben∣Spinosad∣Spiromesifen∣Spirotetramat∣Thiacloprid∣Thiamethoxam∣Trifloxystrobin
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Acetonitrile
Quantity 1 mL
Concentration or Composition (by Analyte or Components) MGK 264 isomer B 7.5 μg/mL∣MGK 264 isomer A 12.5 μg/mL∣Acetamiprid 20 μg/mL∣Azoxystrobin 20 μg/mL∣Bifenazate 20 μg/mL∣Bifenthrin 20 μg/mL∣Carbaryl 20 μg/mL∣Carbofuran 20 μg/mL∣Chlorantraniliprole 20 μg/mL∣Chlorpyrifos 20 μg/mL∣Clofentezine 20 μg/mL∣Diazinon 20 μg/mL∣Dimethoate 20 μg/mL∣Ethoprophos 20 μg/mL∣Etoxazole 20 μg/mL∣Fenoxycarb 20 μg/mL∣Imazalil 20 μg/mL∣Malathion 20 μg/mL∣Metalaxyl 20 μg/mL∣Methiocarb 20 μg/mL∣MGK 264 20 μg/mL∣Myclobutanil 20 μg/mL∣Parathion-methyl 20 μg/mL∣Permethrin 20 μg/mL∣Phosmet 20 μg/mL∣Prallethrin 20 μg/mL∣Propoxur 20 μg/mL∣Pyridaben 20 μg/mL∣Spinosad 20 μg/mL∣Spiromesifen 20 μg/mL∣Spirotetramat 20 μg/mL∣Thiacloprid 20 μg/mL∣Thiamethoxam 20 μg/mL∣Trifloxystrobin 20 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.