Learn More
Oregon Pesticide Mixture 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Product Code. 30310963
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30310963 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30310963 Supplier Dr. Ehrenstorfer Supplier No. DREGA09000473AL
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
Add to basket
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Naled∣Oxamyl∣Paclobutrazol∣Permethrin∣Phosmet∣Piperonyl butoxide∣Prallethrin∣Propiconazole∣Propoxur∣Pyrethrins∣Pyridaben∣Spinosad∣Spiromesifen∣Spirotetramat∣Spiroxamine∣Tebuconazole∣Thiacloprid∣Thiamethoxam∣Trifloxystrobin
CAS 300-76-5, 23135-22-0, 76738-62-0, 52645-53-1, 732-11-6, 51-03-6
Analyte or Component Names Naled∣Oxamyl∣Paclobutrazol∣Permethrin∣Phosmet∣Piperonyl butoxide∣Prallethrin∣Propiconazole∣Propoxur∣Pyrethrins∣Pyridaben∣Spinosad∣Spiromesifen∣Spirotetramat∣Spiroxamine∣Tebuconazole∣Thiacloprid∣Thiamethoxam∣Trifloxystrobin
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Acetonitrile
Quantity 1 mL
Concentration or Composition (by Analyte or Components) Naled 100 μg/mL∣Oxamyl 100 μg/mL∣Paclobutrazol 100 μg/mL∣Permethrin 100 μg/mL∣Phosmet 100 μg/mL∣Piperonyl butoxide 100 μg/mL∣Prallethrin 100 μg/mL∣Propiconazole 100 μg/mL∣Propoxur 100 μg/mL∣Pyrethrins 100 μg/mL∣Pyridaben 100 μg/mL∣Spinosad 100 μg/mL∣Spiromesifen 100 μg/mL∣Spirotetramat 100 μg/mL∣Spiroxamine 100 μg/mL∣Tebuconazole 100 μg/mL∣Thiacloprid 100 μg/mL∣Thiamethoxam 100 μg/mL∣Trifloxystrobin 100 μg/mL