missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Oregon Green™ 488 Maleimide
Description
The thiol reactive Oregon Green™ 488 maleimide can be used to can be used to create green fluorescent bioconjugates with excitation/emission maxima ∼496/524 nm.
This fluorinated analog of fluorescein overcomes some of the key limitations of fluorescein, including greater photostability and a lower pKa (pKa ∼ 4.7 versus 6.4 for fluorescein), making its fluorescence essentially pH insensitive in the physiological pH range.
This fluorinated analog of fluorescein overcomes some of the key limitations of fluorescein, including greater photostability and a lower pKa (pKa ∼ 4.7 versus 6.4 for fluorescein), making its fluorescence essentially pH insensitive in the physiological pH range.
Specifications
Specifications
| Quantity | 5 mg |
| Product Type | Maleimide |
| Content And Storage | Store in freezer (-5 to -30°C) and protect from light. |
| Chemical Reactivity | Thiol |
| Shipping Condition | Room Temperature |
| Reactive Moiety | Maleimide |
| Label or Dye | Oregon Green™ 488 |
| Label Type | Classic Dyes |
| Product Line | Oregon Green |
| Emission | 524 |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?