missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ Oregon Green™ 488 Iodoacetamide, mixed isomers
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Description
The thiol reactive Oregon Green™ 488 iodoacetamide can be used to can be used to create green fluorescent bioconjugates with excitation/emission maxima ∼496/524 nm.
This fluorinated analog of fluorescein overcomes some of the key limitations of fluorescein, including greater photostability and a lower pKa (pKa ∼ 4.7 versus 6.4 for fluorescein), making its fluorescence essentially pH insensitive in the physiological pH range.
This fluorinated analog of fluorescein overcomes some of the key limitations of fluorescein, including greater photostability and a lower pKa (pKa ∼ 4.7 versus 6.4 for fluorescein), making its fluorescence essentially pH insensitive in the physiological pH range.
Specifications
Specifications
| Quantity | 5 mg |
| Product Type | Iodoacetamide |
| Content And Storage | Store in freezer (-5 to -30°C) and protect from light. |
| Chemical Reactivity | Thiol |
| Shipping Condition | Room Temperature |
| Reactive Moiety | Alkyl Halide, Iodoacetamide |
| Label or Dye | Oregon Green™ 488 |
| Label Type | Classic Dyes |
| Product Line | Oregon Green |
| Emission | 524 |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction