Learn More
Hirschmann™ Opus™ Dispenser
Product Code. 11692269
Change view
Click to view available options
Volume (Metric):
10 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
11692269 10 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11692269 Supplier Hirschmann™ Supplier No. 9581010
  • ONLINE EXCLUSIVE PRICE
    3164.36 EUR
    2366.00 EUR / Pack of 1
    Save 798.36 EUR
    25% Off
Estimated Shipment: 10-12-2026
Dispenses directly from the reagent bottle using a motor-powered reagent recirculation system. Hirschmann Instruments™ Opus™ Dispenser provides consistent results through electronic control.
  • Electronic control and dispensing of volumes
  • A light protection window for the valve block is also included in the scope of delivery
  • TFT touchscreen with user-friendly menu
  • Method memory for nine different dispensing procedures with storage of all settings
  • Serial dispensing up to 9999 times
  • Standard stepper function
  • Media recirculating system for simple and rapid venting without reagent loss
  • Free rotation
  • Superior materials and reliable piston sealing, even in the case of problematic media
  • Brown glass cylinder available for light-sensitive media
  • USB and RS232 interface for PC connection
  • Integration in automated procedures possible
Specifications
Accuracy 0.6%
Product Type Electronic Bottletop Dispenser
Volume (Metric) 10 mL
Display TFT Touchscreen
Includes EU, US, UK plug type
Voltage 100/240 V