Learn More
Optima™ LC/MS Solvents Starter Kit, Includes 1L Acetonitrile, 1L Methanol, 1L Ipa And 1L Water Optima™ LC/MS Grade
Product Code. 11988379 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 Box
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11988379 1 Box 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11988379 Supplier Fisher Chemical Supplier No. LCMSKIT
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket
Starter kit with reagents and amber glass bottle for development of the mobile phase for LC/MS
  • The starter kit includes a 1L amber glass bottle for each of the following reagents:
    • A955-1, Optima LC/MS Acetonitrile
    • A456-1. Optima LC/MS Methanol
    • A461-1, Optima LC/MS IPA
    • W6-1, Optima LC/MS Water
  • 1L empty amber glass bottle is included to mix reagents for the mobile phase
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Box
For Use With (Application) Creating Mobile Phase
Product Type LC/MS Solvents Starter Kit