Learn More
ONO-AE3-208, MedChemExpress
Product Code. 16464047
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16464047 50 mg 50mg
16438138 10 mg 10mg
16410598 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16464047 Supplier Medchem Express Supplier No. HY50901
MedChemExpress ONO-AE3-208 is a selective and orally active EP4 receptor antagonist with a Ki of 1.3 nM. ONO-AE3-208 shows less potently affects EP3, FP, and TP receptors (Ki of 30 nM, 790 nM, and 2400 nM, respectively). ONO-AE3-208 suppresses cell invasion, migration, and metastasis of prostate cancer.
ONO-AE3-208 is a selective and orally active EP4 receptor antagonist with a Ki of 1.3 nM. ONO-AE3-208 shows less potently affects EP3, FP, and TP receptors (Ki of 30 nM, 790 nM, and 2400 nM, respectively). ONO-AE3-208 suppresses cell invasion, migration, and metastasis of prostate cancer.
Specifications
Chemical Name or Material ONO-AE3-208
CAS 402473-54-5
Color White
Purity Grade Notes Research
Molecular Formula C24H21FN2O3
Quantity 50 mg
Synonym AE 3-208
Solubility Information DMSO : 33.33 mg/mL (82.41 mM; Need ultrasonic)
SMILES N#CC1=CC=C(C(NC(C(C2=C3C=CC=CC3=C(F)C=C2)C)=O)=C1)CCCC(O)=O
Molecular Weight (g/mol) 404.43
Formula Weight 404.43
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death