Learn More
Invitrogen™ OneComp eBeads™ Compensation Beads
Product Code. 15516296
Change view
Click to view available options
Quantity:
100 tests
25 tests
Unit Size:
100 tests
25 tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15516296 25 tests 25 tests
15242448 100 tests 100 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Contains Sodium azide
Product Code. 15516296 Supplier Invitrogen™ Supplier No. 01111141
  • ONLINE EXCLUSIVE PRICE
    160.00 EUR
    119.65 EUR / 25 tests
    Save 40.35 EUR
    25% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
Contains Sodium azide
Spherical particles that can be stained with individual fluorochrome-conjugated antibodies for use as single-color compensation controls. OneComp eBeads react with antibodies of mouse, rat and hamster origin, and are immunoglobulin light chain-independent. 1 drop (50μL)/test
  • Each drop of beads contains both a positive population that will capture any mouse, rat or hamster antibody and a negative population that will not react with antibody
  • This bimodal distribution can be used for single-color compensation controls in multicolor flow cytometry experiments
  • Cross-reacts to some antibodies of rabbit origin, but have not been extensively tested for this reactivity
  • Designed for use in compensation with all fluorochromes excited by blue (488nm), green (532nm), yellow-green (561nm), and red (633 to 635nm) lasers
  • Compatible with eFluor™ 450 but is not optimized for compensation of other fluorochromes excited by a violet (405nm) laser
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Flow Cytometer
No. of Tests 25 tests
Quantity 25 tests
Product Line eBeads
Product Type Compensation Beads