Learn More
Ondansetron, TRC
Product Code. 30137419
Change view
Click to view available options
Quantity:
10 mg
50 mg
500 mg
Unit Size:
10mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137419 500 mg 500mg
30132420 50 mg 50mg
30012483 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30137419 Supplier Toronto Research Chemicals Supplier No. TRCO655005500MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 99614-02-5
Molecular Formula C18 H19 N3 O
Molecular Weight (g/mol) 293.36
Synonym Ondansetron, 1,2,3,9-Tetrahydro-9-methyl-3-[(2-methyl-1H-imidazol-1-yl)methyl]-4H-carbazol-4-one, 1,2,3,4-Tetrahydro-9-methyl-3-[(2-methyl-1H-imidazol-1-yl)methyl]-9H-carbazol-4-one, GR 38032, GR 38032X, Ondanles, Setron, Yatrox, Zofran, Zophren, Zudan
IUPAC Name 9-methyl-3-[(2-methylimidazol-1-yl)methyl]-2,3-dihydro-1H-carbazol-4-one
SMILES Cc1nccn1CC2CCc3c(C2=O)c4ccccc4n3C
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C18H19N3O/c1-12-19-9-10-21(12)11-13-7-8-16-17(18(13)22)14-5-3-4-6-15(14)20(16)2/h3-6,9-10,13H,7-8,11H2,1-2H3
Formula Weight 293.1528
Percent Purity >95%
Chemical Name or Material Ondansetron
Grade HPLC