Learn More
Fisherbrand™ omniPAGE Mini Vertical Electrophoresis System
Product Code. 12347257 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
No. of Gels:
2 to 4
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Gels Unit Size
12347257 2 to 4 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12347257
  • ONLINE EXCLUSIVE PRICE
    773.00 EUR
    700.91 EUR / Pack of 1
    Save 72.09 EUR
    9% Off
View all promotions
 Restricted Item
Estimated Shipment: 01-03-2027
Suitable for the separation of proteins in polyacrylamide gels.
  • High throughput capability
  • Rapid casting
  • Reversible combs serves as loading indicators which aids pipette-well alignment to prevent sample loading errors
  • Interchangeable modular inserts for slab gels, 2-D electrophoresis and electroblotting
Specifications
Type Vertical Electrophoresis System
Gel Dimensions W x H: 8 x 8.5 cm (3.15 x 3.35 in.)
Includes omniPAGE Mini, 10 x 10cm Dual, 2 sets of glass plates, Bonded spacers (1mm thick), 2 x 12 sample, combs (1mm thick) including caster, External casting upstand
No. of Gels 2 to 4
Volume (English) Buffer 0.06 to 0.32 gal.
Volume (Metric) Buffer 250 to 1200 mL
Dimensions (L x W x H) 13 x 19 x 15 cm ( 5.12 x 7.48 x 5.90 in.)
Weight (English) 3.97 lb.
Weight (Metric) 1.8 kg