missing translation for 'onlineSavingsMsg'
Learn More

Omethoate, TRC

Product Code. 30138538
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138538 1 g 1g
30135505 250 mg 250mg
30041383 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138538 Supplier Toronto Research Chemicals Supplier No. TRCO6355001G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1113-02-6
Molecular Formula C5 H12 N O4 P S
Molecular Weight (g/mol) 213.19
Synonym Phosphorothioic acid, O,O-dimethyl S-[2-(methylamino)-2-oxoethyl] ester, Phosphorothioic acid, O,O-dimethyl S-(methylcarbamoylmethyl) ester (6CI), Phosphorothioic acid, O,O-dimethyl ester, S-ester with 2-mercapto-N-methylacetamide (7CI,8CI), BAY 45432, Bayer 45432, Dimethoate O-analog, Dimethoate PO isologue, Dimethoate oxon, Dimethoate oxygen analog, Dimethoxon, Folimat, Folimat 4E, Le-mat, O,O-Dimethyl S-[(methylcarbamoyl)methyl] phosphorothioate, Omethoate, PO-Dimethoate, Spraymate L 1700
IUPAC Name 2-dimethoxyphosphorylsulfanyl-N-methylacetamide
SMILES CNC(=O)CSP(=O)(OC)OC

Specifications

Quantity 1 g
InChI Formula InChI=1S/C5H12NO4PS/c1-6-5(7)4-12-11(8,9-2)10-3/h4H2,1-3H3,(H,6,7)
Formula Weight 213.0225
Chemical Name or Material Omethoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.