missing translation for 'onlineSavingsMsg'
Learn More

Omeprazole Sulfide, TRC

Product Code. 17961855
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17961855 25 mg 25mg
17912555 10 mg 10mg
17954325 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17961855 Supplier Toronto Research Chemicals Supplier No. TRCO63502025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73590-85-9
Molecular Formula C17 H19 N3 O2 S
Molecular Weight (g/mol) 329.42
Synonym 5-Methoxy-2-[[(4-methoxy-3,5-dimethylpyridin-2-yl)methyl]sulphanyl]-1H-benzimidazole, Esomeprazole Magnesium Trihydrate Imp. C (EP), Omeprazole Imp. C (EP), Ufiprazole
IUPAC Name 5-methoxy-2-[(4-methoxy-3,5-dimethylpyridin-2-yl)methylsulfanyl]-1H-benzimidazole
SMILES COc1ccc2[nH]c(SCc3ncc(C)c(OC)c3C)nc2c1

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C17H19N3O2S/c1-10-8-18-15(11(2)16(10)22-4)9-23-17-19-13-6-5-12(21-3)7-14(13)20-17/h5-8H,9H2,1-4H3,(H,19,20)
Formula Weight 329.1198
Percent Purity >95%
Chemical Name or Material 5-Methoxy-2-([(4-methoxy-3,5-dimethylpyridin-2-yl)methyl]sulphanyl)-1H-benzimidazole (Ufiprazole)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.