Description
Add a wide range of magnifications using Carl Zeiss™ and Olympus™ Objective Lenses for Thermo Scientific™ ArrayScan™ HCA Readers and the Thermo Scientific™ CellInsight™ NXT High Content Screening (HCS) Platform.
The Olympus 2X (0.08NA) PlanApo objective lens is suitable for use with the Thermo Scientific CellInsight CX5, CX7 LED, and CX7 LZR high content screening (HCS) platforms. It has a magnification power suited for subcellular imaging and a numerical aperture that strikes a balance between resolution and versatility.
Specifications
Specifications
| Type | Objective Lens |
| Product Line | Olympus |
| Description | Olympus™ 2X Objective, PlanApo (0.08 NA) |
| For Use With (Equipment) | CellInsight CX5 HCS Platform |
| Lens Type | Dry |
| Magnification | 2x (0.08NA) |
| Quantity | 1 Ea. |