Learn More
Tocris Bioscience™ Oltipraz
GREENER_CHOICE
Nrf2 activator
Brand:  Tocris Bioscience™ 5294/10
Encompass
Product Code. 15746675
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Oltipraz is a Nrf2 activator; activates Nrf, which binds to antioxidant response elements (AREs) and induces Phase-I and -II drug-metabolizing enzymes including CYP2B and NQO1. Also increases mRNA levels of multiple hepatic xenobiotic transporters. Elevates expression of genes encoding anti-oxidant and multidrug resistance-associated proteins. Displays chemopreventive activity.
Specifications
Specifications
4-Methyl-5-(2-pyrazinyl)-3H-1,2-dithiole-3-thione
64224-21-1
10 mg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions