Learn More
Oltipraz, TRC
Product Code. 17932915
Change view
Click to view available options
Quantity:
25 mg
250 mg
500 mg
Unit Size:
250mg
25mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17932915 25 mg 25mg
17980695 250 mg 250mg
17953005 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17932915 Supplier Toronto Research Chemicals Supplier No. TRCO58605025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 64224-21-1
Molecular Formula C8 H6 N2 S3
Molecular Weight (g/mol) 226.34
Synonym 4-Methyl-5-pyrazinyl-3H-1,2-dithiole-3-thione, NSC 347901, RP 35972
IUPAC Name 4-methyl-5-pyrazin-2-yl-dithiole-3-thione
SMILES CC1=C(SSC1=S)c2cnccn2
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C8H6N2S3/c1-5-7(12-13-8(5)11)6-4-9-2-3-10-6/h2-4H,1H3
Formula Weight 225.9693
Chemical Name or Material Oltipraz