Learn More
Oltipraz, MedChemExpress
Product Code. 16465906
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16465906 100 mg 100mg
16451447 50 mg 50mg
16441636 500 mg 500mg
16425238 10 mg 10mg
16401046 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16465906 Supplier Medchem Express Supplier No. HY12519
MedChemExpress Oltipraz has an inhibitory effect on HIF-1α activation in a time-dependent manner, completely abrogating HIF-1α induction at ≥10 μM concentrations, the IC50 of Oltipraz for HIF-1α inhibition is 10 μM. Oltipraz is a potent Nrf2 activator.
Oltipraz has an inhibitory effect on HIF-1α activation in a time-dependent manner, completely abrogating HIF-1α induction at ≥10 μM concentrations, the IC50 of Oltipraz for HIF-1α inhibition is 10 μM. Oltipraz is a potent Nrf2 activator.
Specifications
Chemical Name or Material Oltipraz
Physical Form Solid
CAS 64224-21-1
Color Red
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C8H6N2S3
For Use With (Application) Cancer-programmed cell death
Synonym RP 35972 NSC 347901
Solubility Information DMSO : 6 mg/mL (26.51 mM; Need ultrasonic and warming)
SMILES S=C1SSC(C2=NC=CN=C2)=C1C
Molecular Weight (g/mol) 226.34
Formula Weight 226.34
Percent Purity 99.34%
Grade Research