Learn More
Tocris Bioscience™ Oltipraz
Nrf2 activator
Brand: Tocris Bioscience™ 5294/50
This item is not returnable.
View return policy
Description
Oltipraz is a Nrf2 activator; activates Nrf, which binds to antioxidant response elements (AREs) and induces Phase-I and -II drug-metabolizing enzymes including CYP2B and NQO1. Also increases mRNA levels of multiple hepatic xenobiotic transporters. Elevates expression of genes encoding anti-oxidant and multidrug resistance-associated proteins. Displays chemopreventive activity.Specifications
| 4-Methyl-5-(2-pyrazinyl)-3H-1,2-dithiole-3-thione | |
| 64224-21-1 | |
| 50 mg |