missing translation for 'onlineSavingsMsg'
Learn More

Oleyl Alcohol, Approx. 99%, MP Biomedicals™

Product Code. 11489291 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11489291 500 mg 500mg
11499291 1 g 1g
11409301 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11489291 Supplier MP Biomedicals Supplier No. 0215599690
  • ONLINE EXCLUSIVE PRICE
    102.00 EUR
    96.60 EUR / 500mg
    Save 5.40 EUR
    5% Off
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 143-28-2
Molecular Formula C18H36O
Molecular Weight (g/mol) 268.49
MDL Number MFCD00002993
InChI Key ALSTYHKOOCGGFT-KTKRTIGZSA-N
Synonym oleyl alcohol, cis-9-octadecen-1-ol, z-octadec-9-en-1-ol, ocenol, conditioner 1, dermaffine, lancol, novol, oceol, oleol
PubChem CID 5284499
ChEBI CHEBI:73504
IUPAC Name (9Z)-octadec-9-en-1-ol
SMILES CCCCCCCC\C=C/CCCCCCCCO

Specifications

Color Colorless
Density 0.85g/mL
Assay Percent Range Approx. 99%
Quantity 500 mg
Formula Weight 268.49
Physical Form Liquid
Chemical Name or Material Oleyl Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.