missing translation for 'onlineSavingsMsg'
Learn More

Oleic acid, 71%, MP Biomedicals™

Product Code. 11428120 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 g
1 kg
Unit Size:
1kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11428120 500 g 500g
11438120 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11428120 Supplier MP Biomedicals Supplier No. 0210250090
  • ONLINE EXCLUSIVE PRICE
    105.00 EUR
    99.75 EUR / 500g
    Save 5.25 EUR
    5% Off
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 112-80-1
Molecular Formula C18H34O2
Molecular Weight (g/mol) 282.47
MDL Number MFCD00064242
InChI Key ZQPPMHVWECSIRJ-MDZDMXLPSA-N
Synonym oleic acid, cis-9-octadecenoic acid, cis-oleic acid, elaidoic acid, oleate, glycon wo, wecoline oo, pamolyn 100, glycon ro, z-octadec-9-enoic acid
PubChem CID 445639
ChEBI CHEBI:16196
IUPAC Name (9E)-octadec-9-enoic acid
SMILES CCCCCCCC\C=C\CCCCCCCC(O)=O

Specifications

Color Colorless to White
Density 0.895g/mL
Assay Percent Range 0.71
Quantity 500 g
Formula Weight 282.52
Physical Form Liquid To Solid
Chemical Name or Material Oleic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.