Learn More
3M™ Oil Sorbent Roll, T-150
Product Code. 17138650
Change view
Click to view available options
Length (Metric):
40 m
Unit Size:
1 roll
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
17138650 40 m 1 roll
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17138650 Supplier 3M™ Supplier No. 7000088321
  • ONLINE EXCLUSIVE PRICE
    267.00 EUR
    169.09 EUR / 1 rolls
    Save 97.91 EUR
    36% Off
Estimated Shipment: 04-01-2027
Cut to cover small areas or lay directly upon open waters, shorelines, or discharge streams. In addition, sorbent rolls can be placed on docks, in boats, and under leaking machines.
  • Use to cover large spills, or as a precaution in areas where large spills may occur
  • Ideal for absorbing oil from areas where water may also be present
  • Can be wrung out and re-used many times over
  • Cut to cover small areas or lay on open waters, shorelines, or discharge streams
TRUSTED_SUSTAINABILITY
Specifications
Product Type Sorbent Roll
Color Brown
Diameter (Metric) 480 mm
Format Roll
For Use With (Application) Spill Absorption/Containment
Length (Metric) 40 m
Model T-150