Learn More
OHAUS™ Harvard Junior™ Mechanical Balance
Product Code. 15584729 Shop All Ohaus Products
Change view
Click to view available options
Capacity:
4.4 lb., 2 kg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Unit Size
15584729 4.4 lb., 2 kg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.
Product Code. 15584729 Supplier OHAUS™ Supplier No. 80252050
  • ONLINE EXCLUSIVE PRICE
    191.00 EUR
    141.45 EUR / Pack of 1
    Save 49.55 EUR
    25% Off
In Stock
CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.
Includes
  • Eight-piece metal mass set (2 x 100g, 2 x 50g, 2 x 25g, 2 x 10g)
  • Bowl-type pans for use in weighing round objects
  • Teacher-developed, standards-based activity guide with background information and assessment guide with reproducibles
Unique design combines colorful, plastic two-pan balance with the durability and accuracy of the more expensive Harvard Trip Balances. OHAUS Harvard Junior Mechanical Balance incorporates precision-engineered metal beam and zero-adjust compensator.
  • Interchangeable pans enable a broader range of objects to be weighed
  • Stackable platforms maximize storage space
  • Built-in transportation/storage lock protects the balance's longevity by eliminating oscillation
  • Spring-loaded zero-adjust compensator
  • Manual damping device minimizes oscillation and speeds up weighing
  • Built-in carrying handle for easy transport around a room or from class to class
  • Capacity: 70.5 oz. (2000g)
  • Readability: 0.5g
Warranty
Three years
TRUSTED_SUSTAINABILITY
Specifications
Diameter (English) Outer Pan 6 in.
Diameter (Metric) Outer Pan 15.2 cm
Depth (English) 9.44 in.
Depth (English) Pan 5.9 in.
Depth (Metric) 240.00 mm
Depth (Metric) Pan 15 cm
Height (English) 6.29 in.
Height (Metric) 160.000 mm
Includes Plastic Plates and Bowls
Model HJ2001
Material ABS Plastic, Metal, Stainless Steel
Width (English) 5.51 in.
Width (Metric) 140.00 mm
Warranty 3 Years
Product Line Harvard Junior
Capacity 4.4 lb., 2 kg
Readability 0.500000 g
Description Mechanical Scale, HJ2001
Languages Chinese, English (US), French, German, Italian, Japanese, Korean, Polish, Portuguese (BR), Spanish (MX)
Display Type Analog
Type Harvard Junior™ Mechanical Balance