Learn More
Ofloxacin, MP Biomedicals™
Product Code. 11486971 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11486971 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11486971 Supplier MP Biomedicals Supplier No. 0219901805
Ofloxacin, White Powder
  • It is used to study pneumococcal cell wall-degrading virulence factors.
  • It is a synthetic fluoroquinolone with broad-spectrum antibacterial activity. It is a nalidixic acid analog.
TRUSTED_SUSTAINABILITY
Specifikationer
CAS 82419-36-1
Color White
Loss on Drying 0.0002
Molecular Formula C18H20FN3O4
Quantity 5 g
Synonym Oflocet, Ofloxacine, Oxaldin, Tarivid
Solubility Information Soluble in 1 N NaOH(50mg/mL).
InChI Key GSDSWSVVBLHKDQ-UHFFFAOYNA-N
Optical Rotation +1 to -1°(Lit.)
Molecular Weight (g/mol) 361.37
Percent Purity 0.9952
Physical Form Powder
Visa mer Visa mindre
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.