missing translation for 'onlineSavingsMsg'
Learn More

Ofloxacin, MedChemExpress

Product Code. 16460778
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16460778 5 g 5g
16402059 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16460778 Supplier Medchem Express Supplier No. HYB0125

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ofloxacin (Hoe-280) is a fluoroquinolone whose primary mechanism of action is inhibition of bacterial DNA gyrase. Ofloxacin shows inhibitory activity against vaccinia virus (VV).

Ofloxacin (Hoe-280) is a fluoroquinolone whose primary mechanism of action is inhibition of bacterial DNA gyrase. Ofloxacin shows inhibitory activity against vaccinia virus (VV).

Specifications

Chemical Name or Material Ofloxacin
Physical Form Powder
CAS 82419-36-1
Color White
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C18H20FN3O4
For Use With (Application) COVID-19-immunoregulation
Synonym Hoe-280
Solubility Information DMSO : ≥ 4 mg/mL (11.07 mM)
SMILES O=C(C(C1=O)=CN2C(C)COC3=C(N4CCN(C)CC4)C(F)=CC1=C23)O
Molecular Weight (g/mol) 361.37
Formula Weight 361.37
Percent Purity 98.97%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.