missing translation for 'onlineSavingsMsg'
Learn More

Octyl sulfate sodium salt, Sigma-Aldrich™

Product Code. 32238467
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32238467 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 32238467 Supplier Sigma Aldrich Supplier No. 1122680500
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

For surfactant tests

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Octyl sulfate sodium salt
CAS 142-31-4
Melting Point 195°C (decomposition)
Quantity 500 g
Synonym Octyl sulfate sodium salt, Sodium octyl sulfate, Octyl sulfate sodium salt
Molecular Weight (g/mol) 232.27
Grade Not Available
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.