missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Octreotide acetate

Product Code. 16483317
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16483317 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16483317 Supplier MedChemExpress Supplier No. HY17365

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Octreotide acetate, a long-acting synthetic analog of native somatostatin, inhibits growth hormone, glucagon, and insulin more potently.

Specifications

Purity 98.95%
CAS 79517-01-4
Content And Storage Sealed storage, away from moisture and light∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture and light)
For Use With (Application) COVID-19-immunoregulation
Molecular Weight (g/mol) 1079.29 g/mol
Product Type Peptides
Quantity 50 mg
Shipping Condition Room temperature
Solubility DMSO : ≥ 29 mg/mL (26.87 mM) ∣H2O : 25 mg/mL (23.16 mM∣Need ultrasonic)
Synonym SMS 201-995 acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.