missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Octenyltrichlorosilane, tech.

Product Code. 15400248
Change view
Click to view available options
Quantity:
25g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15400248 25g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15400248 Supplier Alfa Aesar™ Supplier No. H53434.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Applications
Multiwalled carbon nanotubes functionalized with 7-octenyltrichlorosilane and n-octyltrichlorosilane. A novel fabrication method of organic multilayer film is by using 7-octenyltrichlorosilane. Octenyltrichlorosilane is used as functionalizing agent of the raw, purified and functionalized nanotubes.

Solubility
Reacts with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
TRUSTED_SUSTAINABILITY

Specifications

CAS 153447-97-3
Quantity 25g
Molecular Formula C8H15Cl3Si
Linear Formula CH2=CH(CH2)6SiCl3
MDL Number MFCD00039292
UN Number UN2987
Flash Point 94°C (201°F)
Synonym 7-octenyltrichlorosilane, silane, trichloro-7-octenyl, trichloro-7-octenylsilane, trichloro oct-7-en-1-yl silane, silane, trichloro-7-octen-1-yl, 7-oct-1-enyltrichlorosilane, octenyltrichlorosilane, mixture of isomers, 7-octenyl trichlorosilane, trichloro oct-7-enyl silane
InChI Key MFISPHKHJHQREG-UHFFFAOYSA-N
SMILES C=CCCCCCC[Si](Cl)(Cl)Cl
IUPAC Name trichloro(oct-7-enyl)silane
Molecular Weight (g/mol) 245.643
PubChem CID 104122
Formula Weight 245.65
Grade Technical
Chemical Name or Material Octenyltrichlorosilane
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.