Learn More
Octenidine dihydrochloride, 98%
Product Code. 15428808
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 15428808
Brand: Thermo Scientific Alfa Aesar H62257.03
Request Bulk or Custom Format
CAS: 70775-75-6 | C36H64Cl2N4
Octenidine dihydrochloride is used as a antiseptic for skin, mucous Membranes and wounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Octenidine dihydrochloride is used as a antiseptic for skin, mucous Membranes and wounds.
Solubility
Soluble in dimethyl sulfoxide and methanol.
Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 70775-75-6
Molecular Formula C36H64Cl2N4
MDL Number MFCD01938808
Synonym N,N'-[Decane-1,10-diyldi-1(4H)-pyridyl-4-ylidene]bis(octylammonium) dichloride
Specifications
Melting Point 214°C
Quantity 1 g
Merck Index 14,6754
Solubility Information Soluble in dimethyl sulfoxide and methanol.
Formula Weight 623.83
Percent Purity 98%
Chemical Name or Material Octenidine dihydrochloride
compliance-icons
Product Identifier
  • Octenidine dihydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Octenidine dihydrochloride