CAS RN 70775-75-6
CAS RN 70775-75-6
IUPAC Name:
dihydrogen N-(1-{10-[4-(octylimino)-1,4-dihydropyridin-1-yl]decyl}-1,4-dihydropyridin-4-ylidene)octan-1-amine dichloride
Synonyms:
octenidine hcl
Molecular Weight (g/mol):
623.84
Molecular Formula:
C36H64Cl2N4
InChi Key:
SMGTYJPMKXNQFY-UHFFFAOYSA-N
SMILES:
[H+].[H+].[Cl-].[Cl-].CCCCCCCCN=C1C=CN(CCCCCCCCCCN2C=CC(C=C2)=NCCCCCCCC)C=C1
1
–
2
of
2
results
1
Octenidine dihydrochloride, 98%
CAS: 70775-75-6 Molecular Formula: C36H64Cl2N4 MDL Number: MFCD01938808 Synonym: N,N'-[Decane-1,10-diyldi-1(4H)-pyridyl-4-ylidene]bis(octylammonium) dichloride
2
Octenidine dihydrochloride, MedChemExpress
MedChemExpress Octenidine dihydrochloride is an effective antiseptic compound for skin mucous membranes and wounds.