missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Octanoic acid, Sigma-Aldrich™
Specifications
Specifications
| Chemical Name or Material | Octanoic acid |
| CAS | 124-07-2 |
| Melting Point | 16°C |
| Density | 0.91 g/cm³ at 20°C |
| Boiling Point | 237°C (1013 hPa) |
| Flash Point | > 110°C (closed cup) |
| Molecular Formula | CH3(CH2)6COOH |
| Quantity | 2.5 L |
| Synonym | Octanoic acid, Octanoic acid |
| Molecular Weight (g/mol) | 144.21 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?