Learn More
Octanedioic Acid, Sigma-Aldrich™
Product Code. 15810631
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15810631 500 g 500g
15800631 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15810631 Supplier Sigma Aldrich Supplier No. 8188000500
 Restricted Item
Estimated Shipment: 25-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 505-48-6
Specifications
Melting Point 139°C to 142°C
Boiling Point 229°C to 231°C/20 hPa
Molecular Formula HOOC(CH2)6COOH
MDL Number MFCD00004428
Quantity 500 g
Synonym Octanedioic acid, Suberic acid, Octanedioic acid
Molecular Weight (g/mol) 174.19
Grade Research