Product Code. 15625450
Shop All Solstice Research Chemicals Products
Click to view available options
Quantity:
100 mL
500 mL
1 L
Unit Size:
1 L
100mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
Quantity
Packaging
Unit Size
15625450
1 L
Glass Bottle
1 L
15615450
100 mL
Glass Bottle
100mL
15635450
500 mL
Glass Bottle
500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC. We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code.
15625450
Supplier
Honeywell Research Chemicals
Supplier No.
4122361L
This item is currently unavailable or has been discontinued. View the product page for possible alternatives.
View alternative products
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC. We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Reagent Grade, 98%
≥95% for general laboratory use.
Chemical Identifiers
CAS
111-65-9
Molecular Formula
C8H18
Molecular Weight (g/mol)
114.232
MDL Number
MFCD00009556
InChI Key
TVMXDCGIABBOFY-UHFFFAOYSA-N
Synonym
n-octane, oktan, oktanen, ottani, oktanen dutch, heptane, methyl, oktan polish, ottani italian, octane, all isomers, unii-x1rv0b2fjv
PubChem CID
356
ChEBI
CHEBI:17590
IUPAC Name
octane
SMILES
CCCCCCCC
Specifications
Melting Point
-57°C
Density
0.703g/mL at 25°C
Boiling Point
125°C to 127°C
Flash Point
13°C (55.4°F)
Infrared Spectrum
Conforms to structure
Packaging
Glass Bottle
Linear Formula
CH3 (CH2 )6 CH3
Refractive Index
n20/D 1.398
Quantity
1 L
UN Number
UN1262
Beilstein
1696875
Vapor Density
3.9 (vs air)
Formula Weight
114.23g/mol
Percent Purity
98%
Physical Form
Liquid
Chemical Name or Material
Octane
Show More
Show Less
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our
Privacy Policy .
Spot an opportunity for improvement?