Learn More
Octane, ≥99% (GC), Honeywell™ Riedel-de-Haën™
Product Code. 15697030
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15697030 100 mL Glass bottle 100mL
15607040 500 mL Glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.
We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code. 15697030 Supplier Honeywell Riedel-de Haën Supplier No. 74821100ML
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.
We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Puriss. p.a.,≥99.0% (GC)
  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 111-65-9
Molecular Formula C8H18
Molecular Weight (g/mol) 114.232
MDL Number MFCD00009556
InChI Key TVMXDCGIABBOFY-UHFFFAOYSA-N
Synonym n-octane, oktan, oktanen, ottani, oktanen dutch, heptane, methyl, oktan polish, ottani italian, octane, all isomers, unii-x1rv0b2fjv
PubChem CID 356
ChEBI CHEBI:17590
IUPAC Name octane
SMILES CCCCCCCC
Specifications
Melting Point -57°C
Density 0.703g/mL at 25°C
Boiling Point 125°C to 127°C
Flash Point 13°C (55.4°F)
Packaging Glass bottle
Linear Formula CH3(CH2)6CH3
Refractive Index 1.398
Quantity 100 mL
UN Number UN1262
Beilstein 1696875
Vapor Density 3.9 (vs air)
Formula Weight 114.23g/mol
Percent Purity ≥99.0% (GC)
Chemical Name or Material Octane
Mehr anzeigen Weniger anzeigen