missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Octane-1-sulfonic acid sodium salt, LiChropur™ EMD-Millipore
Specifications
Specifications
| CAS | 5324-84-5 |
| Density | 345kg/m3 |
| Packaging | Glass |
| Molecular Formula | C8H17NaO3S |
| MDL Number | MFCD00007544 |
| Quantity | 25 g |
| Synonym | sodium 1-octanesulfonate, sodium octane-1-sulfonate, 1-octanesulfonic acid sodium salt, sodium octanesulfonate, 1-octanesulfonic acid, sodium salt, 1-octanesulfonate, sodium, sodium octylsulfonate, unii-j46ez5jyb3, octyl sodium sulfonate, ipc-alks-8 |
| InChI Key | HRQDCDQDOPSGBR-UHFFFAOYSA-M |
| SMILES | [Na+].CCCCCCCCS([O-])(=O)=O |
| IUPAC Name | sodium octane-1-sulfonate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?